Potassium tris(trifluoromethanesulfonyl)methanide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
P2520-1G
P2520-5G
TCP2520-1GEA
206.02
USD
TCP2520-1G
TCP2520-5G
Potassium tris(trifluoromethanesulfonyl)methanide ≥98.0% (by titrimetric analysis)
Potassium tris(trifluoromethanesulfonyl)methanide
Formula:
C₄F₉KO₆S₃ |
MDL Number:
MFCD18072174 CAS Number: 114395-69-6 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...