3-Bromo-9-(3,5-dimethylphenyl)-9H-carbazole ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
B5621-1G
B5621-200MG
TCB5621-1GEA
245
USD
TCB5621-1G
TCB5621-200MG
3-Bromo-9-(3,5-dimethylphenyl)-9H-carbazole ≥98.0% (by HPLC)
3-Bromo-9-(3,5-dimethylphenyl)-9H-carbazole
Formula:
C₂₀H₁₆BrN MW: 350.26 g/mol |
CAS Number:
1141017-77-7 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 113.0 to 117.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...