2,7-Bis[N,N-bis(4-methoxyphenyl)amino]-9,9-spirobi[9H-fluorene] ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
B4959-1G
B4959-200MG
76219-464EA
772
USD
76219-464
76219-466
2,7-Bis[N,N-bis(4-methoxyphenyl)amino]-9,9-spirobi[9H-fluorene] ≥98.0% (by HPLC, total nitrogen)
2,7-Bis[N,N-bis(4-methoxyphenyl)amino]-9,9-spirobi[9H-fluorene]
Formula:
C₅₃H₄₂N₂O₄ |
CAS Number:
1138220-69-5 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 184.0 to 188.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...