1,4-Di(4-pyridyl)benzene ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 4,4'-(1,4-Phenylene)bispyridine, 1,4-Di(pyridin-4-yl)benzene

P1550-1G P1550-200MG
TCP1550-1GEA 374.5 USD
TCP1550-1G TCP1550-200MG
1,4-Di(4-pyridyl)benzene ≥95.0% (by GC)
1,4-Di(4-pyridyl)benzene
Formula: C₁₆H₁₂N₂
MW: 232.29 g/mol
MDL Number: MFCD00445845
CAS Number: 113682-56-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR