2-Phenylpyridine 1-oxide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

P2775-1G P2775-250MG
TCP2775-1GEA 203 USD
TCP2775-1G TCP2775-250MG
2-Phenylpyridine 1-oxide ≥98.0% (by HPLC, titration analysis)
2-Phenylpyridine 1-oxide
Formula: C₁₁H₉NO
MDL Number: MFCD00234705
CAS Number: 1131-33-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 156.0 to 160.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR