Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-tetradecanoyl-L-lysine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
Fmoc-Lys(myristoyl)-OH, Fmoc-Lys(Myr)-OH, Nalpha-Fmoc-Nepsilon-tetradecanoyl-L-lysine
F1143-1G
F1143-5G
TCF1143-1GEA
176
USD
TCF1143-1G
TCF1143-5G
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-tetradecanoyl-L-lysine ≥98.0% (by HPLC, total nitrogen)
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-tetradecanoyl-L-lysine
Formula:
C₃₅H₅₀N₂O₅ MW: 578.79 g/mol |
CAS Number:
1128181-23-6 |
Specification Test Results
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 138.0 to 142.0 deg-C |
Specific rotation [a]20/D | +13.0 to +17.0 deg (C=1, CHCL3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...