Ethyl 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylic acid Ethyl ester, Ethyl 1-cyclopropyl-6,7-difluoro-8-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylate

E1147-1G E1147-5G
TCE1147-1GEA 29 USD
TCE1147-1G TCE1147-5G
Ethyl 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylate ≥98.0% (by GC)
Ethyl 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-8-methoxy-4-oxo-3-quinolinecarboxylate
Formula: C₁₆H₁₅F₂NO₄
MW: 323.30 g/mol
Storage Temperature: Freezer
MDL Number: MFCD05864419
CAS Number: 112811-71-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 183.0 to 187.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR