4-Allylpyrocatechol ≥98.0% (by GC)

Supplier: TCI America
Warning

A2947-100MG A2947-10MG
TCA2947-100MGEA 685 USD
TCA2947-100MG TCA2947-10MG
4-Allylpyrocatechol ≥98.0% (by GC)
4-Allylpyrocatechol
Formula: C₉H₁₀O₂
Boiling Pt: 123°C
Storage Temperature: Refrigerator
MDL Number: MFCD01548379
CAS Number: 1126-61-0

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR