7,7-Dimethyl-7H-benzo[c]fluorene ≥94.0% (by GC)
Supplier: TCI America
D5125-1G
D5125-5G
TCD5125-1GEA
121
USD
TCD5125-1G
TCD5125-5G
7,7-Dimethyl-7H-benzo[c]fluorene ≥94.0% (by GC)
7,7-Dimethyl-7H-benzo[c]fluorene
Formula:
C₁₉H₁₆ MW: 244.34 g/mol |
CAS Number:
112486-09-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 94.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...