Glyceryl Ascorbate ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2-O-(2,3-Dihydroxypropyl)-L-ascorbic acid
G0451-25G
G0451-5G
TCG0451-25GEA
307.6
USD
TCG0451-25G
TCG0451-5G
Glyceryl Ascorbate ≥95.0% (by titrimetric analysis)
Glyceryl Ascorbate
Formula:
C₉H₁₄O₈ MW: 250.20 g/mol |
CAS Number:
1120360-13-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 156.0 to 161.0 deg-C |
Specific rotation [a]20/D | +52.0 to +57.0 deg (C=1, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...