3,3''-Di(9H-carbazol-9-yl)-1,1':3',1''-terphenyl ≥97.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
3,3''-Di(9H-carbazol-9-yl)-m-terphenyl
D4904-1G
D4904-200MG
TCD4904-1GEA
372
USD
TCD4904-1G
TCD4904-200MG
3,3''-Di(9H-carbazol-9-yl)-1,1':3',1''-terphenyl ≥97.0% (by HPLC, total nitrogen)
3,3''-Di(9H-carbazol-9-yl)-1,1':3',1''-terphenyl
Formula:
C₄₂H₂₈N₂ MW: 560.70 g/mol |
CAS Number:
1116499-73-0 |
Specification Test Results
Appearance | White to Light gray to Light yellow powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...