[(1H-Benzotriazol-1-yl)methyl]triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
B4486-1G
B4486-5G
TCB4486-1GEA
55
USD
TCB4486-1G
TCB4486-5G
[(1H-Benzotriazol-1-yl)methyl]triphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
[(1H-Benzotriazol-1-yl)methyl]triphenylphosphonium chloride
Formula:
C₂₅H₂₁ClN₃P |
MDL Number:
MFCD00960175 CAS Number: 111198-09-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...