3,6-Dibromo-9-heptyl-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

D5404-1G D5404-5G
TCD5404-1GEA 156 USD
TCD5404-1G TCD5404-5G
3,6-Dibromo-9-heptyl-9H-carbazole ≥98.0% (by GC)
3,6-Dibromo-9-heptyl-9H-carbazole
Formula: C₁₉H₂₁Br₂N
MW: 423.19 g/mol
MDL Number: MFCD31618132
CAS Number: 1103535-99-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR