2,5,8,11,14-Pentaoxaheptadec-16-yne ≥98.0% (by GC)

Supplier: TCI America

P2249-100MG P2249-25MG
76220-396EA 181 USD
76220-396 76220-398
2,5,8,11,14-Pentaoxaheptadec-16-yne ≥98.0% (by GC)
2,5,8,11,14-Pentaoxaheptadec-16-yne
Formula: C₁₂H₂₂O₅
Storage Temperature: Refrigerator
CAS Number: 1101668-39-6

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR