3-Bromo-4-ethoxybenzaldehyde ≥97.0% (by GC)
Supplier: TCI America
|
Warning
|
B5079-1G
B5079-5G
TCB5079-1GEA
46
USD
TCB5079-1G
TCB5079-5G
3-Bromo-4-ethoxybenzaldehyde ≥97.0% (by GC)
3-Bromo-4-ethoxybenzaldehyde
Formula:
C₉H₉BrO₂ |
MDL Number:
MFCD00672941 CAS Number: 108373-05-3 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 70.0 to 74.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...