2-Methylquinoline N-Oxide ≥98.0% (by GC)

Supplier: TCI America

M3000-1G M3000-5G
TCM3000-1GEA 65 USD
TCM3000-1G TCM3000-5G
2-Methylquinoline N-Oxide ≥98.0% (by GC)
2-Methylquinoline N-Oxide
Formula: C₁₀H₉NO
MW: 159.19 g/mol
CAS Number: 1076-28-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR