Tetradecamethylcycloheptasiloxane ≥95.0% (by GC)

Supplier: TCI America
Warning

T2678-1ML T2678-5ML
TCT2678-1MLEA 381.46 USD
TCT2678-1ML TCT2678-5ML
Tetradecamethylcycloheptasiloxane ≥95.0% (by GC)
Tetradecamethylcycloheptasiloxane
Formula: C₁₄H₄₂O₇Si₇
MW: 519.08 g/mol
Boiling Pt: 151°C
Melting Pt: -32°C
CAS Number: 107-50-6
EINECS: 203-496-9

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR