2-(3,5-Dibromophenyl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

D4902-1G D4902-200MG
TCD4902-1GEA 227 USD
TCD4902-1G TCD4902-200MG
2-(3,5-Dibromophenyl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2-(3,5-Dibromophenyl)-4,6-diphenyl-1,3,5-triazine
Formula: C₂₁H₁₃Br₂N₃
MDL Number: MFCD25562933
CAS Number: 1073062-59-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 244.0 to 248.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR