2-(2-Hydroxy-4-methoxyphenyl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Warning
|
Synonyms:
2-(4,6-Diphenyl-1,3,5-triazin-2-yl)-5-methoxyphenol
H1531-25G
H1531-5G
TCH1531-25GEA
362
USD
TCH1531-25G
TCH1531-5G
2-(2-Hydroxy-4-methoxyphenyl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2-(2-Hydroxy-4-methoxyphenyl)-4,6-diphenyl-1,3,5-triazine
Formula:
C₂₂H₁₇N₃O₂ MW: 355.40 g/mol |
MDL Number:
MFCD23102325 CAS Number: 106556-36-9 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 206.0 to 210.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...