Sertindole ≥95.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 1-(2-(4-(5-Chloro-1-(4-fluorophenyl)-1H-indol-3-yl)piperidin-1-yl)ethyl)imidazolidin-2-one, 1-[2-[4-[5-Chloro-1-(4-fluorophenyl)-1H-indol-3-yl]-1-piperidinyl]ethyl]-2-imidazolidinone

S0942-10MG S0942-50MG
TCS0942-10MGEA 137 USD
TCS0942-10MG TCS0942-50MG
Sertindole ≥95.0% (by HPLC)
Sertindole
Formula: C₂₄H₂₆ClFN₄O
Melting Pt: 155°C
Storage Temperature: Refrigerator
MDL Number: MFCD00867749
CAS Number: 106516-24-9
Merck Index: 14,08466

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 12.00 to 13.00 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR