Rufinamide ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 1-(2,6-Difluorobenzyl)-1H-1,2,3-triazole-4-carboxamide

R0143-100MG R0143-1G R0143-25MG
TCR0143-100MGEA 43 USD
TCR0143-100MG TCR0143-1G TCR0143-25MG
Rufinamide ≥98.0% (by HPLC)
Rufinamide
Formula: C₁₀H₈F₂N₄O
MW: 238.20 g/mol
MDL Number: MFCD00865314
CAS Number: 106308-44-5
Merck Index: 14,08293

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 22.50 to 24.50 %
Melting point 239.0 to 243.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR