2,8-Diiododibenzothiophene ≥97.0% (by GC)

Supplier: TCI America

D4870-1G D4870-200MG
TCD4870-1GEA 488.6 USD
TCD4870-1G TCD4870-200MG
2,8-Diiododibenzothiophene ≥97.0% (by GC)
2,8-Diiododibenzothiophene
Formula: C₁₂H₆I₂S
MDL Number: MFCD00275724
CAS Number: 105404-91-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 223.0 to 227.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR