Dimethyldivinylsilane ≥98.0% (by GC)

Supplier: TCI America
Danger

D4866-25ML D4866-5ML
TCD4866-25MLEA 114 USD
TCD4866-25ML TCD4866-5ML
Dimethyldivinylsilane ≥98.0% (by GC)
Dimethyldivinylsilane
Formula: C₆H₁₂Si
Boiling Pt: 82°C
Flash Pt: –22°C
MDL Number: MFCD00053855
CAS Number: 10519-87-6
EINECS: 234-060-6
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR