N-Carbobenzoxy-1,10-diaminodecane hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America
Warning

C3471-1G C3471-5G
TCC3471-1GEA 121 USD
TCC3471-1G TCC3471-5G
N-Carbobenzoxy-1,10-diaminodecane hydrochloride ≥98.0% (by HPLC)
N-Carbobenzoxy-1,10-diaminodecane hydrochloride
Formula: C₁₈H₃₀N₂O₂·HCl
MDL Number: MFCD08272290
CAS Number: 1051420-13-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 175.0 to 179.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR