4,8-Bis(dodecyloxy)benzo[1,2-b:4,5-b']dithiophene ≥98.0% (by HPLC)

Supplier: TCI America

B5454-1G B5454-200MG
TCB5454-1GEA 493 USD
TCB5454-1G TCB5454-200MG
4,8-Bis(dodecyloxy)benzo[1,2-b:4,5-b']dithiophene ≥98.0% (by HPLC)
4,8-Bis(dodecyloxy)benzo[1,2-b:4,5-b']dithiophene
Formula: C₃₄H₅₄O₂S₂
MW: 558.92 g/mol
CAS Number: 1044795-04-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR