Diethyl (Ethoxymethyl)phosphonate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
(Ethoxymethyl)phosphonic acid Diethyl ester
D5095-25G
D5095-5G
TCD5095-25GEA
352
USD
TCD5095-25G
TCD5095-5G
Diethyl (Ethoxymethyl)phosphonate ≥98.0% (by GC)
Diethyl (Ethoxymethyl)phosphonate
Formula:
C₇H₁₇O₄P MW: 196.18 g/mol Boiling Pt: 103°C |
CAS Number:
10419-80-4 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...