Trospium chloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: rel-(1R,3r,5S)-3-(2-Hydroxy-2,2-diphenylacetoxy)spiro[bicyclo[3.2.1]octane-8,1'-pyrrolidin]-1'-ium chloride

T3305-1G T3305-200MG
TCT3305-1GEA 207.43 USD
TCT3305-1G TCT3305-200MG
Trospium chloride ≥98.0% (by HPLC, total nitrogen)
Trospium chloride
Formula: C₂₅H₃₀ClNO₃
MW: 427.97 g/mol
MDL Number: MFCD00865254
CAS Number: 10405-02-4
EINECS: 233-875-4
Merck Index: 14,09786

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR