2-Bromo-3-(2-ethylhexyl)-5-iodothiophene ≥97.0% (by GC) stabilized

Supplier: TCI America


Stabilization: Stabilized with Copper chip

B4585-1G B4585-5G
TCB4585-1GEA 113 USD
TCB4585-1G TCB4585-5G
2-Bromo-3-(2-ethylhexyl)-5-iodothiophene ≥97.0% (by GC) stabilized
2-Bromo-3-(2-ethylhexyl)-5-iodothiophene
Formula: C₁₂H₁₈BrIS
Storage Temperature: Freezer
CAS Number: 1034352-30-1

Order Now

Specification Test Results

Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR