[RuCl(p-cymene)((R)-tolbinap)]Cl
Supplier: TCI America
Synonyms:
Chloro[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride
R0148-1G
R0148-200MG
TCR0148-1GEA
169.5
USD
TCR0148-1G
TCR0148-200MG
[RuCl(p-cymene)((R)-tolbinap)]Cl
[RuCl(p-cymene)((R)-tolbinap)]Cl
Formula:
C₅₈H₅₄Cl₂P₂Ru MW: 984.99 g/mol |
CAS Number:
1034001-51-8 |
Specification Test Results
Appearance | Light yellow to Brown powder to crystal |
IR | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...