2,7-Bis[N-(m-tolyl)anilino]-9,9'-spirobi[9H-fluorene] ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 2,7-Bis[phenyl(m-tolyl)amino]-9,9'-spirobi[9H-fluorene]

B4882-1G B4882-200MG
TCB4882-1GEA 465.5 USD
TCB4882-1G TCB4882-200MG
2,7-Bis[N-(m-tolyl)anilino]-9,9'-spirobi[9H-fluorene] ≥98.0% (by HPLC, total nitrogen)
2,7-Bis[N-(m-tolyl)anilino]-9,9'-spirobi[9H-fluorene]
Formula: C₅₁H₃₈N₂
MW: 678.88 g/mol
MDL Number: MFCD11112143
CAS Number: 1033035-83-4

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 211.0 to 215.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR