2-Bromo-4-(tert-butyl)aniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B5325-25G B5325-5G
TCB5325-25GEA 228 USD
TCB5325-25G TCB5325-5G
2-Bromo-4-(tert-butyl)aniline ≥98.0% (by GC, titration analysis)
2-Bromo-4-(tert-butyl)aniline
Formula: C₁₀H₁₄BrN
MW: 228.13 g/mol
Boiling Pt: 265°C
MDL Number: MFCD00052693
CAS Number: 103273-01-4

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR