3-Phenyl-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

P2401-1G P2401-5G
TCP2401-1GEA 88 USD
TCP2401-1G TCP2401-5G
3-Phenyl-9H-carbazole ≥98.0% (by GC)
3-Phenyl-9H-carbazole
Formula: C₁₈H₁₃N
MDL Number: MFCD22052094
CAS Number: 103012-26-6

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 220.0 to 224.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR