Triethoxy[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]silane ≥98.0% (by GC)

Supplier: TCI America

T3585-25ML T3585-5ML
TCT3585-25MLEA 44 USD
TCT3585-25ML TCT3585-5ML
Triethoxy[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]silane ≥98.0% (by GC)
Triethoxy[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]silane
Formula: C₁₄H₂₈O₄Si
Boiling Pt: 130°C
MDL Number: MFCD00069159
CAS Number: 10217-34-2
EINECS: 425-050-4

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific gravity (20/20) 1.0050 to 1.0150


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR