(3-Bromophenyl)diphenylphosphine oxide ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B5530-1G
B5530-200MG
76218-840EA
182
USD
76218-840
76218-842
(3-Bromophenyl)diphenylphosphine oxide ≥98.0% (by GC)
(3-Bromophenyl)diphenylphosphine oxide
Formula:
C₁₈H₁₄BrOP MW: 357.19 g/mol |
MDL Number:
MFCD30533951 CAS Number: 10212-04-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...