1-Methylimidazole hydrobromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
M3212-25G
M3212-5G
TCM3212-25GEA
204
USD
TCM3212-25G
TCM3212-5G
1-Methylimidazole hydrobromide ≥98.0% (by HPLC, titration analysis)
1-Methylimidazole hydrobromide
Formula:
C₄H₆N₂·HBr |
MDL Number:
MFCD18251295 CAS Number: 101023-58-9 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...