Imidazole hydrobromide, low water content ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

I1006-1G I1006-5G
TCI1006-1GEA 84.42 USD
TCI1006-1G TCI1006-5G
Imidazole hydrobromide, low water content ≥98.0% (by HPLC, titration analysis)
Imidazole hydrobromide
Formula: C₃H₄N₂·HBr
Melting Pt: 221°C
CAS Number: 101023-55-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Water max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR