Trimethyl(4-vinylphenyl)silane ≥95.0% (by GC) stabilized

Supplier: TCI America

Synonyms: 1-Ethenyl-4-(trimethylsilyl)benzene, 4-Trimethylsilylstyrene

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

T3919-1G T3919-250MG
TCT3919-1GEA 146 USD
TCT3919-1G TCT3919-250MG
Trimethyl(4-vinylphenyl)silane ≥95.0% (by GC) stabilized
Trimethyl(4-vinylphenyl)silane
Formula: C₁₁H₁₆Si
MW: 176.33 g/mol
Boiling Pt: 90°C
Storage Temperature: Refrigerator
MDL Number: MFCD05663945
CAS Number: 1009-43-4

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %
Specific gravity (20/20) 0.8880 to 0.9020

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR