Sodium bis(fluorosulfonyl)imide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

S0992-1G S0992-5G
TCS0992-1GEA 178 USD
TCS0992-1G TCS0992-5G
Sodium bis(fluorosulfonyl)imide ≥98.0% (by titrimetric analysis)
Sodium bis(fluorosulfonyl)imide
Formula: F₂NNaO₄S₂
MDL Number: MFCD29905041
CAS Number: 100669-96-3
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR