5-Phenylisoxazole ≥98.0% (by GC)

Supplier: TCI America

P2309-1G P2309-5G
TCP2309-1GEA 94 USD
TCP2309-1G TCP2309-5G
5-Phenylisoxazole ≥98.0% (by GC)
5-Phenylisoxazole
Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 260°C
Melting Pt: 22°C
MDL Number: MFCD00090038
CAS Number: 1006-67-3

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR