1-(Azidomethyl)pyrene ≥98.0% (by HPLC)

Supplier: TCI America

A2791-1G A2791-200MG
TCA2791-1GEA 658 USD
TCA2791-1G TCA2791-200MG
1-(Azidomethyl)pyrene ≥98.0% (by HPLC)
1-(Azidomethyl)pyrene
Formula: C₁₇H₁₁N₃
MW: 257.30 g/mol
Storage Temperature: Refrigerator
CAS Number: 1006061-57-9

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 15.50 to 17.00 %
Melting point 66.0 to 70.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR