9,9-Diphenyl-9H-9-silafluorene ≥98.0% (by GC)
Supplier: TCI America
D5034-1G
D5034-200MG
TCD5034-1GEA
301
USD
TCD5034-1G
TCD5034-200MG
9,9-Diphenyl-9H-9-silafluorene ≥98.0% (by GC)
9,9-Diphenyl-9H-9-silafluorene
Formula:
C₂₄H₁₈Si |
CAS Number:
5550-08-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 146.0 to 151.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...