p-Xylylenediphosphonic acid ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

Synonyms: (1,4-Phenylenebis(methylene))diphosphonic acid

X0074-1G X0074-5G
TCX0074-1GEA 165 USD
TCX0074-1G TCX0074-5G
p-Xylylenediphosphonic acid ≥97.0% (by HPLC, titration analysis)
p-Xylylenediphosphonic acid
Formula: C₈H₁₂O₆P₂
MW: 266.13 g/mol
Melting Pt: 280°C
MDL Number: MFCD05663703
CAS Number: 4546-06-9
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystaline
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR