1,2-Benzenedisulfonic imide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

B4139-1G B4139-5G
TCB4139-1GEA 241 USD
TCB4139-1G TCB4139-5G
1,2-Benzenedisulfonic imide ≥98.0% (by HPLC, titration analysis)
1,2-Benzenedisulfonic imide
Formula: C₆H₅NO₄S₂
MDL Number: MFCD07779379
CAS Number: 4482-01-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 194.0 to 198.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR