Methyl 4-(Phenylcarbamoyl)benzoate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
4-(Phenylcarbamoyl)benzoic acid methyl ester, Methyl 4-(phenylcarbamoyl)benzoate
M2637-1G
M2637-5G
TCM2637-1GEA
50
USD
TCM2637-1G
TCM2637-5G
Methyl 4-(Phenylcarbamoyl)benzoate ≥98.0% (by GC)
Methyl 4-(Phenylcarbamoyl)benzoate
Formula:
C₁₅H₁₃NO₃ MW: 255.27 g/mol |
MDL Number:
MFCD00157832 CAS Number: 3814-10-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 192.0 to 196.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...