9-Methyl-9H-pyrido[3,4-b]indole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M2818-1G M2818-5G
TCM2818-1GEA 104.26 USD
TCM2818-1G TCM2818-5G
9-Methyl-9H-pyrido[3,4-b]indole ≥98.0% (by GC, titration analysis)
9-Methyl-9H-pyrido[3,4-b]indole
Formula: C₁₂H₁₀N₂
MDL Number: MFCD18803860
CAS Number: 2521-07-5

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 105.0 to 109.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR