Octadecyl rhodamine B chloride

Supplier: TCI America
Warning

Synonyms: 6-(Diethylamino)-N,N-diethyl-9-[2-[(octadecyloxy)carbonyl]phenyl]-3H-xanthen-3-iminium chloride, 3,6-Bis(diethylamino)-9-[2-[(octadecyloxy)carbonyl]phenyl]xanthylium chloride, N-[6-(Diethylamino)-9-[2-[(octadecyloxy)carbonyl]phenyl]-3H-xanthen-3-ylidene]-N-ethylethanaminium chloride

O0512-10MG O0512-50MG
TCO0512-10MGEA 238 USD
TCO0512-10MG TCO0512-50MG
Octadecyl rhodamine B chloride
Octadecyl rhodamine B chloride
Formula: C₄₆H₆₇ClN₂O₃
MW: 731.50 g/mol
MDL Number: MFCD00467892
CAS Number: 65603-19-2

Order Now

Specification Test Results

Appearance Orange to Amber to Dark red powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) gt. 3.5 to 4.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR