OAC1 ≥98.0% (by GC)

Supplier: TCI America

O0490-10MG O0490-50MG
TCO0490-10MGEA 135 USD
TCO0490-10MG TCO0490-50MG
OAC1 ≥98.0% (by GC)
OAC1
Formula: C₁₄H₁₁N₃O
Storage Temperature: Freezer
MDL Number: MFCD00572806
CAS Number: 300586-90-7

Order Now

Specification Test Results

Appearance White to Green to Brown powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR