Formamidine hydroiodide ≥99.0% (by titrimetric analysis) for perovskite precursor

Supplier: TCI America

Synonyms: FAI, Formamidinium iodide

F1263-1G F1263-25G F1263-5G
TCF1263-1GEA 90 USD
TCF1263-1G TCF1263-25G TCF1263-5G
Formamidine hydroiodide ≥99.0% (by titrimetric analysis) for perovskite precursor
Formamidine hydroiodide
Formula: CH₄N₂·HI
MW: 171.97 g/mol
MDL Number: MFCD28369273
CAS Number: 879643-71-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (Argentmetric Titration) min. 99.0 %
Iodine content min. 99.0 %
Purity (Trace metal basis) min. 99.99 %
Solubility in 2-Propanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR