Repaglinide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Ethoxy-4-[2-[[(1S)-3-methyl-1-[2-(1-piperidinyl)phenyl]butyl]amino]-2-oxoethyl]benzoic acid

R0179-1G R0179-200MG
TCR0179-1GEA 330 USD
TCR0179-1G TCR0179-200MG
Repaglinide ≥98.0% (by HPLC, titration analysis)
Repaglinide
Formula: C₂₇H₃₆N₂O₄
MW: 452.59 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00906179
CAS Number: 135062-02-1
Merck Index: 14,08136

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 131.0 to 135.0 deg-C
Specific rotation [a]20/D +7.0 to +10.0 deg (C=1,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR