Aprepitant ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 5-(((2R,3S)-2-((R)-1-(3,5-Bis(trifluoromethyl)phenyl)ethoxy)-3-(4-fluorophenyl)morpholino)methyl)-1H-1,2,4-triazol-3(2H)-one

A3135-100MG A3135-500MG
76219-306EA 188 USD
76219-306 76219-308
Aprepitant ≥98.0% (by HPLC)
Aprepitant
Formula: C₂₃H₂₁F₇N₄O₃
Storage Temperature: Refrigerator
MDL Number: MFCD08277635
CAS Number: 170729-80-3
Merck Index: 14,00753

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 250.0 to 255.0 deg-C
Specific rotation [a]20/D +67.0 to +71.0 deg (C=1,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR